siRNA Details
| Gene Symbol: LOC101928495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110409.1 | 3 | 494 | |||
| GenBank mRNA | BC013926.1 | - | 3 | 504 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE564886.1 | - | 478 | Chr. - - [-] on Build | ||

LOC101928495
Genomic Map