siRNA Details
| Gene Symbol: SNORA17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054908.1 | - | 6 | 2343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNORA17B| Gene Symbol: SNORA17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054908.1 | - | 6 | 2343 | ||