siRNA Details
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 6 | 1080 | ||
NR_126398.1 | 8 | 1262 | ||||
NR_126399.1 | 6 | 1071 | ||||
GenBank mRNA | AK094450.1 | - | 6 | 1028 | ||
BC120993.2 | - | 6 | 1006 |
