siRNA Details
| Gene Symbol: LINC01208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109968.1 | 3 | 376 | |||
| GenBank mRNA | BX094192.1 | - | 3 | 376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA029727.1 | - | 3 | 376 | Chr. 3 - 176604302 - 176635532 [-] on Build GRCh38 | |

LINC01208
Genomic Map