siRNA Details
| Gene Symbol: ERICH3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121670.1 | 3 | 494 | |||
| NR_121671.1 | 3 | 401 | ||||
| GenBank mRNA | BX092187.1 | - | 3 | 494 | ||
| CR627203.1 | - | 3 | 401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH3-AS1| Gene Symbol: ERICH3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121670.1 | 3 | 494 | |||
| NR_121671.1 | 3 | 401 | ||||
| GenBank mRNA | BX092187.1 | - | 3 | 494 | ||
| CR627203.1 | - | 3 | 401 | |||