siRNA Details
| Gene Symbol: LAMA5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109922.1 | 3 | 588 | |||
| NR_109923.1 | 2 | 248 | ||||
| NR_109924.1 | 3 | 461 | ||||
| GenBank mRNA | BQ188912.1 | - | 2 | 247 | ||
| CD675432.1 | - | 3 | 459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAMA5-AS1| Gene Symbol: LAMA5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109922.1 | 3 | 588 | |||
| NR_109923.1 | 2 | 248 | ||||
| NR_109924.1 | 3 | 461 | ||||
| GenBank mRNA | BQ188912.1 | - | 2 | 247 | ||
| CD675432.1 | - | 3 | 459 | |||