siRNA Details
| Gene Symbol: IL21R-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037158.1 | 3 | 2472 | |||
| GenBank mRNA | AK093371.1 | - | 3 | 2474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL21R-AS1| Gene Symbol: IL21R-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037158.1 | 3 | 2472 | |||
| GenBank mRNA | AK093371.1 | - | 3 | 2474 | ||