siRNA Details
| Gene Symbol: HP09025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109783.1 | 2 | 746 | |||
| GenBank mRNA | AB593148.1 | - | 2 | 689 | ||
| BC008248.1 | - | 2 | 746 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HP09025| Gene Symbol: HP09025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109783.1 | 2 | 746 | |||
| GenBank mRNA | AB593148.1 | - | 2 | 689 | ||
| BC008248.1 | - | 2 | 746 | |||