siRNA Details
Gene Symbol: LINC00324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026951.1 | 2 | 742 | |||
GenBank mRNA | AK092109.1 | - | 2 | 742 | ||
BC063002.1 | - | 1 | 300 | |||
BC089396.1 | - | 1 | 271 | |||
BC113564.1 | - | 2 | 583 |
