siRNA Details
| Gene Symbol: LINC01143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126384.1 | 3 | 395 | |||
| GenBank mRNA | BX088774.1 | - | 3 | 395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01143| Gene Symbol: LINC01143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126384.1 | 3 | 395 | |||
| GenBank mRNA | BX088774.1 | - | 3 | 395 | ||