siRNA Details
| Gene Symbol: LOC100133077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121583.1 | 1 | 124 | |||
| GenBank mRNA | AK128414.1 | - | 1 | 124 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100133077| Gene Symbol: LOC100133077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121583.1 | 1 | 124 | |||
| GenBank mRNA | AK128414.1 | - | 1 | 124 | ||