siRNA Details
Gene Symbol: LOC101929237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125433.1 | 3 | 1412 | |||
GenBank mRNA | AK124682.1 | - | 3 | 1400 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125433.1 | 3 | 1412 | |||
GenBank mRNA | AK124682.1 | - | 3 | 1400 |