siRNA Details
Gene Symbol: LOC101929237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125433.1 | 3 | 623 | |||
GenBank mRNA | AK124682.1 | - | 3 | 611 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA318894.1 | - | 3 | 469 | Chr. 8 - 22878142 - 22884420 [+] on Build GRCh38 |
