siRNA Details
| Gene Symbol: C5orf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131245.1 | 3 | 637 | |||
| GenBank mRNA | AK092155.1 | - | 3 | 637 | ||
| BQ950913.1 | - | 3 | 597 | |||
| DQ323997.1 | - | 3 | 412 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf17| Gene Symbol: C5orf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131245.1 | 3 | 637 | |||
| GenBank mRNA | AK092155.1 | - | 3 | 637 | ||
| BQ950913.1 | - | 3 | 597 | |||
| DQ323997.1 | - | 3 | 412 | |||