siRNA Details
| Gene Symbol: LOC105369192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134236.1 | 5 | 984 | |||
| NR_134237.1 | 4 | 908 | ||||
| GenBank mRNA | BC027846.1 | - | 5 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369192| Gene Symbol: LOC105369192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134236.1 | 5 | 984 | |||
| NR_134237.1 | 4 | 908 | ||||
| GenBank mRNA | BC027846.1 | - | 5 | 972 | ||