siRNA Details
Gene Symbol: LOC101927267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120449.1 | 2, 3 | 270 | |||
GenBank mRNA | AK128056.1 | - | 2, 3 | 270 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120449.1 | 2, 3 | 270 | |||
GenBank mRNA | AK128056.1 | - | 2, 3 | 270 |