siRNA Details
Gene Symbol: LINC00898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033377.1 | 4 | 2663 | |||
GenBank mRNA | AK128136.1 | - | 4 | 2660 | ||
BC132762.1 | - | 4 | 996 | |||
BC132788.1 | - | 4 | 996 | |||
BC144501.1 | - | 3 | 915 | |||
BC144502.1 | - | 3 | 1044 |
