siRNA Details
| Gene Symbol: LOC105379178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086294.1 | - | 2 | 507 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105379178| Gene Symbol: LOC105379178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086294.1 | - | 2 | 507 | ||