siRNA Details
| Gene Symbol: LINC01445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120515.1 | 3 | 755 | |||
| NR_120516.1 | 3 | 288 | ||||
| GenBank mRNA | AV709400.1 | - | 3 | 217 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX111274.1 | - | 3 | 288 | Chr. 7 - 54330697 - 54349641 [+] on Build GRCh38 | |

LINC01445
Genomic Map