siRNA Details
Gene Symbol: LOC101928324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125952.1 | 3 | 638 | |||
NR_125953.1 | 2 | 517 | ||||
GenBank mRNA | AL050085.1 | - | 2 | 517 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125952.1 | 3 | 638 | |||
NR_125953.1 | 2 | 517 | ||||
GenBank mRNA | AL050085.1 | - | 2 | 517 |