siRNA Details
| Gene Symbol: LHFPL3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027374.1 | 4 | 624 | |||
| GenBank mRNA | BC050337.1 | - | 4 | 624 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI762394.1 | - | 5 | 720 | Chr. 7 - 104895808 - 104926530 [-] on Build GRCh38 | |

LHFPL3-AS2
Genomic Map