siRNA Details
| Gene Symbol: KCCAT211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110826.1 | 3 | 1363 | |||
| GenBank mRNA | AK056252.1 | - | 3 | 1364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCCAT211| Gene Symbol: KCCAT211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110826.1 | 3 | 1363 | |||
| GenBank mRNA | AK056252.1 | - | 3 | 1364 | ||