siRNA Details
| Gene Symbol: LOC101929269 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134308.1 | 2 | 519 | |||
| GenBank mRNA | BC038544.1 | - | 2 | 519 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI834352.1 | - | 518 | Chr. - - [-] on Build | ||

LOC101929269
Genomic Map