siRNA Details
| Gene Symbol: MYLK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038266.2 | 3 | 780 | |||
| NR_121654.1 | 4 | 687 | ||||
| GenBank mRNA | AV744989.1 | - | 2 | 554 | ||
| BC042384.1 | - | 3 | 736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYLK-AS1| Gene Symbol: MYLK-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038266.2 | 3 | 780 | |||
| NR_121654.1 | 4 | 687 | ||||
| GenBank mRNA | AV744989.1 | - | 2 | 554 | ||
| BC042384.1 | - | 3 | 736 | |||