siRNA Details
| Gene Symbol: KCNAB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046618.2 | 3 | 459 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNAB1-AS1| Gene Symbol: KCNAB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046618.2 | 3 | 459 | |||