siRNA Details
| Gene Symbol: LINC00540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103810.1 | 2 | 1430 | |||
| GenBank mRNA | AK054845.1 | - | 2 | 1697 | ||
| BC035104.2 | - | 2 | 1430 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00540| Gene Symbol: LINC00540 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103810.1 | 2 | 1430 | |||
| GenBank mRNA | AK054845.1 | - | 2 | 1697 | ||
| BC035104.2 | - | 2 | 1430 | |||