siRNA Details
Gene Symbol: LINC00540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103810.1 | 2 | 1986 | |||
GenBank mRNA | AK054845.1 | - | 2 | 2253 | ||
BC035104.2 | - | 2 | 1986 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103810.1 | 2 | 1986 | |||
GenBank mRNA | AK054845.1 | - | 2 | 2253 | ||
BC035104.2 | - | 2 | 1986 |