siRNA Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG057763.1 | - | 1 | 149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR4A1| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG057763.1 | - | 1 | 149 | ||