siRNA Details
| Gene Symbol: LOC101929164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110746.1 | 3 | 564 | |||
| GenBank mRNA | AL133086.1 | - | 3 | 564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929164| Gene Symbol: LOC101929164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110746.1 | 3 | 564 | |||
| GenBank mRNA | AL133086.1 | - | 3 | 564 | ||