siRNA Details
| Gene Symbol: TRIM55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005251316.4 | XP_005251373.1 | 2 | 356 | ||
| XM_011517613.2 | XP_011515915.1 | 2 | 356 | |||
| XM_011517614.2 | XP_011515916.1 | 2 | 279 | |||
| XM_011517615.2 | XP_011515917.1 | 2 | 356 | |||
| XM_011517616.2 | XP_011515918.1 | 2 | 356 | |||
| XM_011517617.2 | XP_011515919.1 | 2 | 477 | |||

TRIM55
Genomic Map