siRNA Details
| Gene Symbol: LOC101929897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125823.1 | 1 | 222 | |||
| GenBank mRNA | DB446853.1 | - | 1 | 173 | ||
| HY042442.1 | - | 1 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929897| Gene Symbol: LOC101929897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125823.1 | 1 | 222 | |||
| GenBank mRNA | DB446853.1 | - | 1 | 173 | ||
| HY042442.1 | - | 1 | 223 | |||