siRNA Details
| Gene Symbol: LINC01166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120626.1 | 2 | 1695 | |||
| NR_120627.1 | 2 | 1529 | ||||
| GenBank mRNA | AK095503.1 | - | 2 | 1529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01166| Gene Symbol: LINC01166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120626.1 | 2 | 1695 | |||
| NR_120627.1 | 2 | 1529 | ||||
| GenBank mRNA | AK095503.1 | - | 2 | 1529 | ||