siRNA Details
Gene Symbol: LOC101927164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110668.1 | 2 | 384 | |||
GenBank mRNA | BC039513.1 | - | 2 | 384 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM465960.1 | - | 2 | 382 | Chr. 1 - 224219615 - 224227574 [+] on Build GRCh38 |
