siRNA Details
| Gene Symbol: LOC101927139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_123718.1 | 3 | 317 | |||
| GenBank mRNA | BX109047.1 | - | 3 | 317 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU853878.1 | - | 3 | 424 | Chr. 1 - 66042424 - 66050339 [-] on Build GRCh38 | |

LOC101927139
Genomic Map