siRNA Details
| Gene Symbol: LINC01266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110118.1 | 4 | 3740 | |||
| GenBank mRNA | AK126307.1 | - | 5 | 3838 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01266| Gene Symbol: LINC01266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110118.1 | 4 | 3740 | |||
| GenBank mRNA | AK126307.1 | - | 5 | 3838 | ||