siRNA Details
| Gene Symbol: LINC00605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033938.1 | 3 | 1163 | |||
| GenBank mRNA | BC035396.1 | - | 3 | 1154 | ||
| BX161461.1 | - | 3 | 1163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00605| Gene Symbol: LINC00605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033938.1 | 3 | 1163 | |||
| GenBank mRNA | BC035396.1 | - | 3 | 1154 | ||
| BX161461.1 | - | 3 | 1163 | |||