siRNA Details
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 599 | |||
GenBank mRNA | BC036758.2 | - | 7 | 610 | 5563071 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126435.1 | 7 | 599 | |||
GenBank mRNA | BC036758.2 | - | 7 | 610 | 5563071 |