siRNA Details
Gene Symbol: LOC441178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117092.1 | 4 | 1432 | |||
GenBank mRNA | AK056600.1 | - | 4 | 1432 | ||
AL832737.1 | - | 6 | 2764 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC441178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117092.1 | 4 | 1432 | |||
GenBank mRNA | AK056600.1 | - | 4 | 1432 | ||
AL832737.1 | - | 6 | 2764 |