siRNA Details
| Gene Symbol: LOC101928600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109904.1 | 4 | 439 | |||
| GenBank mRNA | AK309731.1 | - | 4 | 439 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DC372088.1 | - | 4 | 439 | Chr. 5 - 58542209 - 58558243 [-] on Build GRCh38 | |

LOC101928600
Genomic Map