siRNA Details
| Gene Symbol: LOC101928600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109904.1 | 4 | 624 | |||
| GenBank mRNA | AK309731.1 | - | 4 | 624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928600| Gene Symbol: LOC101928600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109904.1 | 4 | 624 | |||
| GenBank mRNA | AK309731.1 | - | 4 | 624 | ||