siRNA Details
| Gene Symbol: FAM13A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002806.2 | 3 | 722 | |||
| GenBank mRNA | AK093650.1 | - | 1 | 518 | ||
| AK126217.1 | - | 3 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM13A-AS1| Gene Symbol: FAM13A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002806.2 | 3 | 722 | |||
| GenBank mRNA | AK093650.1 | - | 1 | 518 | ||
| AK126217.1 | - | 3 | 722 | |||