siRNA Details
| Gene Symbol: LINC00623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024510.2 | 2 | 2074 | |||
| NR_024511.2 | 3 | 1640 | ||||
| NR_024584.2 | 4 | 1591 | ||||
| NR_109754.2 | 3 | 1480 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | F06803.1 | - | 44 | Chr. - - [-] on Build | ||

LINC00623
Genomic Map