siRNA Details
| Gene Symbol: LOC105373876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135525.1 | 1 | 167 | |||
| GenBank mRNA | BC031974.1 | - | 1 | 172 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373876| Gene Symbol: LOC105373876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135525.1 | 1 | 167 | |||
| GenBank mRNA | BC031974.1 | - | 1 | 172 | ||