siRNA Details
| Gene Symbol: ZNF582-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037159.1 | 4 | 1231 | |||
| NR_037160.1 | 2 | 1234 | ||||
| GenBank mRNA | BC024249.1 | - | 2 | 1168 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF582-AS1| Gene Symbol: ZNF582-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037159.1 | 4 | 1231 | |||
| NR_037160.1 | 2 | 1234 | ||||
| GenBank mRNA | BC024249.1 | - | 2 | 1168 | ||