siRNA Details
Gene Symbol: LOC105378470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134331.1 | 2 | 317 | |||
GenBank mRNA | BX098586.1 | - | 2 | 286 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105378470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134331.1 | 2 | 317 | |||
GenBank mRNA | BX098586.1 | - | 2 | 286 |