siRNA Details
Gene Symbol: LOC106660606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132988.1 | 1 | 68 | |||
GenBank mRNA | BM766743.1 | - | 1 | 68 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM766675.1 | - | 1 | 68 | Chr. 16 - 2268663 - 2273124 [+] on Build GRCh38 |
