siRNA Details
Gene Symbol: LOC105374546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134673.1 | 2 | 462 | |||
NR_134674.1 | 2 | 293 | ||||
NR_134675.1 | 2 | 310 | ||||
GenBank mRNA | BU942553.1 | - | 2 | 278 | ||
BX100535.1 | - | 2 | 462 | |||
DB507158.1 | - | 2 | 310 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU951742.1 | - | 2 | 415 | Chr. 4 - 26859766 - 26860550 [-] on Build GRCh38 |
