siRNA Details
| Gene Symbol: DNM1P35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024595.3 | 5 | 1945 | |||
| GenBank mRNA | AK091244.1 | - | 3 | 1566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNM1P35| Gene Symbol: DNM1P35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024595.3 | 5 | 1945 | |||
| GenBank mRNA | AK091244.1 | - | 3 | 1566 | ||