siRNA Details
Gene Symbol: LOC101927179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125880.1 | 3 | 373 | |||
NR_125881.1 | 3 | 344 | ||||
GenBank mRNA | AL599247.1 | - | 3 | 345 | ||
BX280044.1 | - | 3 | 335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | W58114.1 | - | 336 | Chr. - - [-] on Build |
