siRNA Details
| Gene Symbol: LINC00977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033916.1 | 3 | 451 | |||
| GenBank mRNA | BX090073.1 | - | 3 | 451 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000509893.2 | - | 3 | 461 | Chr. 8 - 129216467 - 129241250 [-] on Build GRCh38 | |

LINC00977
Genomic Map